4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS
4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS
4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS
Thermo Scientific Chemicals

4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS

CAS: 5808-22-0 | C10H6O8S2 | 318.27 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 022414.14
または、製品番号022414-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5808-22-0
IUPAC Name2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
Molecular FormulaC10H10Na2O10S2
InChI KeyQUEAKWJKJBFNEG-UHFFFAOYSA-L
SMILESO.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]
さらに表示
Appearance (Color)White to grey, beige or brown
CommentSensitivity to formaldehyde P.T.
CommentSensitivity to nitrate P.T.
Solution TestP.T.
FormPowder
4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, is used to improve the visualization of trichothecenes on developed TLC plates. It serves as an intermediate in the production of dyes, pigments, chelating reagents, sulfonic and sulfinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only