Chlorobenzene, Spectrophotometric Grade, 99.9%
Chlorobenzene, Spectrophotometric Grade, 99.9%
Chlorobenzene, Spectrophotometric Grade, 99.9%
Thermo Scientific Chemicals

Chlorobenzene, Spectrophotometric Grade, 99.9%

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
数量:
500 mL
1 L
4 L
4 x 1 L
製品番号(カタログ番号) 022921.M1
または、製品番号022921-M1
価格(JPY)
-
数量:
4 x 1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
さらに表示
Optical Absorbance at350nm: ≤0.02AU [US]
Optical Absorbance at290nm: ≥50% transmittance [UK]
Optical Absorbance at295nm: ≥80% transmittance [UK]
Optical Absorbance at370-400nm: ≥99% transmittance [UK]
Optical Absorbance at400nm: ≤0.005AU
さらに表示
Chlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only