1-Propanol, HPLC Grade, 99% min
1-Propanol, HPLC Grade, 99% min
1-Propanol, HPLC Grade, 99% min
Thermo Scientific Chemicals

1-Propanol, HPLC Grade, 99% min

CAS: 71-23-8 | C3H8O | 60.10 g/mol
数量:
1 L
4 L
4 x 1 L
製品番号(カタログ番号) 022932.M1
または、製品番号022932-M1
価格(JPY)
-
数量:
4 x 1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
さらに表示
CommentSub-micron filtered (U.S. specification)
Water Content (Karl Fischer Titration)≤0.08% (U.S. specification)
Optical Absorbance at210nm: ≤1.0 AU (U.S. specification)
Optical Absorbance at210nm: ≥10% Transmittance (non-U.S. specification)
Optical Absorbance at225nm: ≤0.50 AU (U.S. specification)
さらに表示
1-Propanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only