Cobalt(II) iodide, anhydrous, 99.5% (metals basis)
Cobalt(II) iodide, anhydrous, 99.5% (metals basis)
Cobalt(II) iodide, anhydrous, 99.5% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) iodide, anhydrous, 99.5% (metals basis)

Moisture and humidity indicator, catalyst | CAS: 15238-00-3
製品番号(カタログ番号) 023119.14
または、製品番号023119-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15238-00-3
IUPAC Nameλ²-cobalt(2+) diiodide
Molecular FormulaCoI2
InChI KeyAVWLPUQJODERGA-UHFFFAOYSA-L
SMILES[Co++].[I-].[I-]
さらに表示
Total Metal Impurities0.5% max.
Cobalt(II) iodide is used as a moisture and humidity indicator. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only