Cobalt(II) acetate, anhydrous, Co 32% min.
Cobalt(II) acetate, anhydrous, Co 32% min.
Cobalt(II) acetate, anhydrous, Co 32% min.
Cobalt(II) acetate, anhydrous, Co 32% min.
Thermo Scientific Chemicals

Cobalt(II) acetate, anhydrous, Co 32% min.

CAS: 71-48-7 | C4H6CoO4 | 177.02 g/mol
製品番号(カタログ番号) 023138.14
または、製品番号023138-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS71-48-7
IUPAC Nameλ²-cobalt(2+) diacetate
Molecular FormulaC4H6CoO4
InChI KeyQAHREYKOYSIQPH-UHFFFAOYSA-L
SMILES[Co++].CC([O-])=O.CC([O-])=O
さらに表示
Assay (unspecified)96% min
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only