Peroxide, Quantofix™ Test Strips
Peroxide, Quantofix™ Test Strips
Thermo Scientific Chemicals
Peroxide, Quantofix™ Test Strips
製品番号(カタログ番号)数量
023284.QA
または、製品番号023284-QA
1 Kit
製品番号(カタログ番号) 023284.QA
または、製品番号023284-QA
価格(JPY)
-
見積もりを依頼する
数量:
1 Kit
一括またはカスタム形式をリクエストする
仕様
濃度Measures 1 to 25ppm peroxide in ether
数量1 Kit
推奨保存方法Ambient temperatures
タイプPeroxide Test Strip
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
How many strips come in the Peroxide, Quantofix Test Strips (Cat. No. 023284.QA, 023284-QA)?
Peroxide, Quantofix Test Strips comes with a tube of 100 test strips.