Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Actual product may vary
Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Thermo Scientific™

Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers

Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호멸균마개 색상라벨팩당 수
030007
03 0007으로도 사용됨
SterileWhiteTabbed100/Pk., Pre-assembled
030181
03 0181으로도 사용됨
Non-sterileWhiteNo Label500/Pk., Bagged Separately
030193
03 0193으로도 사용됨
Non-sterileYellowNo Label100/Pk., Pre-assembled
030195
03 0195으로도 사용됨
Non-sterileOrangeNo Label500/Pk., Bagged Separately
카탈로그 번호 030007
03 0007으로도 사용됨
제품 가격(KRW)
-
멸균:
Sterile
마개 색상:
White
라벨:
Tabbed
팩당 수:
100/Pk., Pre-assembled

Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Available Options:

  • Nonsterile or gamma irradiation sterilized
  • Labled or non-labled
  • Bulk or individually packed
사양
용량(미터법)60 mL
마개 색상White
마개 크기48 mm
마개 유형Bio-Tite Lid
설명Cap and container assembled, sterile, tabbed label, white caps
직경(미터법)48 mm
라벨Tabbed
케이스당 수500/Cs.
팩당 수100/Pk., Pre-assembled
Closure IncludedYes
물질Virgin Polypropylene
제품 유형Specimen Container
형상Cylindrical
멸균Sterile
Unit SizeCase of 500