Iron(II) bromide, anhydrous, 98% min
Iron(II) bromide, anhydrous, 98% min
Iron(II) bromide, anhydrous, 98% min
Thermo Scientific Chemicals

Iron(II) bromide, anhydrous, 98% min

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 031154.22
または、製品番号031154-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7789-46-0
IUPAC Nameλ²-iron(2+) dibromide
Molecular FormulaBr2Fe
InChI KeyGYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES[Fe++].[Br-].[Br-]
さらに表示
Assay (unspecified)97.6% min
Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only