n-Decane, 99+%
n-Decane, 99+%
n-Decane, 99+%
Thermo Scientific Chemicals

n-Decane, 99+%

CAS: 124-18-5 | C10H22 | 142.286 g/mol
数量:
10 g
100 g
500 g
製品番号(カタログ番号) 031209.36
または、製品番号031209-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS124-18-5
IUPAC Namedecane
Molecular FormulaC10H22
InChI KeyDIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILESCCCCCCCCCC
さらに表示
Appearance (Color)Colorless [EU]
FormClear liquid [EU & US]
Assay (unspecified)≤C-10 carbon: ≤0.5% [US]
Assay (GC)≥99.0% [EU & US]
Refractive Index1.4100-1.4130 @ 20?C [EU]
さらに表示
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only