2-Furaldehyde, ACS, 98% min
2-Furaldehyde, ACS, 98% min
2-Furaldehyde, ACS, 98% min
Thermo Scientific Chemicals

2-Furaldehyde, ACS, 98% min

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol
製品番号(カタログ番号) 031305.A1
または、製品番号031305-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-01-1
IUPAC Namefuran-2-carbaldehyde
Molecular FormulaC5H4O2
InChI KeyHYBBIBNJHNGZAN-UHFFFAOYSA-N
SMILESO=CC1=CC=CO1
さらに表示
Assay (GC)≥98.0%
Free acid (titration)≤0.2meq/g
Residue on Evaporation≤0.5%
It is primarily used in the production of furan derivatives such as furan and furfuryl alcohol. Another major application of furfural is as extraction solvent in refineries. It is also used to accelerate vulcanization, as an ingredient of phenolic resins (Durite) and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only