Chloroform, ACS, 99.8+%
Chloroform, ACS, 99.8+%
Chloroform, ACS, 99.8+%
Thermo Scientific Chemicals

Chloroform, ACS, 99.8+%

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
数量:
1 L
4 L
4 x 4 L
製品番号(カタログ番号) 032614.K7
または、製品番号032614-K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
さらに表示
Impurity contentAcid and chlorine: Passes test
Impurity contentFree chlorine (Cl): Passes test
Impurity contentLead (Pb): ≤0.05ppm
Impurity contentSubstances darkened by sulfuric acid: Passes test
Residue on Evaporation≤0.001%
さらに表示
Chloroform is suitable for pesticide analysis, environmental and GC analysis. It is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform plays an essential role in the RNA isolation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only