Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals - FAQs
Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals - FAQs
View additional product information for Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals - FAQs (033213.22, 033213.36, 033213.A3)
1 product FAQs found
What is used for a stabilizer in Phenol, ACS, 99+%, stab.?
The stabilizer in Phenol, ACS, 99+%, stab. is hypophosphorous acid. It is included at a maximum concentration of 0.15%.