Naphthalene, 99%
Naphthalene, 99%
Naphthalene, 99%
Thermo Scientific Chemicals

Naphthalene, 99%

Formerly important in dyestuff manufacture, as a precursor to phthalic anhydride | CAS: 91-20-3 | C10H8 | 128.17 g/mol
数量:
50 g
500 g
2 kg
製品番号(カタログ番号) 033347.18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-20-3
IUPAC Namenaphthalene
Molecular FormulaC10H8
InChI KeyUFWIBTONFRDIAS-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=C2C=C1
さらに表示
CommentPurchased in the U.S. and in other countries
Appearance (Color)White
FormCrystalline powder or flakes
Color (APHA)≤50 (U.S. specification)
Assay (GC)≥98.5% (non-U.S. specification)
さらに表示
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only