Thermo Scientific Chemicals

Sodium peroxide, ACS, 93% min, Thermo Scientific Chemicals

Catalog number: 033384.22
100 g, Each
Thermo Scientific Chemicals

Sodium peroxide, ACS, 93% min, Thermo Scientific Chemicals

Catalog number: 033384.22
100 g, Each
Quantity
Catalog number: 033384.22
also known as 033384-22
Price (USD)
Price: 124.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1313-60-6
IUPAC Name
disodium dioxidanediide
Molecular Formula
Na2O2
InChI Key
PFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][O-]
Impurity content
Iron, Fe: ≤50ppm
Impurity content
Phosphate PO₄: ≤5ppm
Impurity content
Sulfate SO₄: ≤10ppm
Heavy Metals (as Pb)
≤20ppm
Form
Powder and granules

Description

As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent.Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text