Thermo Scientific Chemicals

Sodium pyrophosphate decahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 033385.A3
2 kg, Each
Thermo Scientific Chemicals

Sodium pyrophosphate decahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 033385.A3
2 kg, Each
Quantity
Catalog number: 033385.A3
also known as 033385-A3
Price (USD)
Price: 219.00
Online price: 186.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13472-36-1
IUPAC Name
tetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular Formula
H20Na4O17P2
InChI Key
VZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Impurity content
SO₄ 0.005%
Insoluble material
0.01%
pH
of a 5% solution 9.5-10.5 at 25?
Trace Metals
(as Pb) 0.001%
Assay (unspecified)
99.0-103.0% Na₄O₇P₂.1₀H₂O

Description

Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text