Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, ACS, 99.0-103.0%

CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol
数量:
100 g
500 g
2.5 kg
製品番号(カタログ番号) 033399.A4
または、製品番号033399-A4
価格(JPY)
-
見積もりを依頼する
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7446-20-0
IUPAC Namezinc(2+) heptahydrate sulfate
Molecular FormulaH14O11SZn
InChI KeyRZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
さらに表示
Assay (unspecified)99.0-103.0% O₄SZn.7H₂O
Impurity contentFe 0.001%
Impurity contentMg 0.005%
Impurity contentPb 0.003%
Impurity contentCa 0.005%
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only