Trimethylgermanium chloride
Trimethylgermanium chloride
Trimethylgermanium chloride
Thermo Scientific Chemicals

Trimethylgermanium chloride

CAS: 1529-47-1 | C3H9ClGe | 153.185 g/mol
製品番号(カタログ番号) 034129.06
または、製品番号034129-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1529-47-1
Molecular FormulaC3H11ClGe
InChI KeyTWBCUFVBXMWAQL-UHFFFAOYSA-N
SMILESCl.C[GeH](C)C
Molecular Weight (g/mol)155.20
FormLiquid
Appearance (Color)Colorless
Assay (unspecified)≥97.0%
Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only