Aluminum chloride, 6.0N Standardized Solution
Aluminum chloride, 6.0N Standardized Solution
Aluminum chloride, 6.0N Standardized Solution
Thermo Scientific Chemicals

Aluminum chloride, 6.0N Standardized Solution

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
数量:
100 mL
250 mL
製品番号(カタログ番号) 035568.AK
または、製品番号035568-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS7446-70-0
化学物質名または材質Aluminum chloride
DOT情報Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION
式量133.34
健康被害1H314-H335
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only