Acetic acid, 1% v/v aq. soln.
Acetic acid, 1% v/v aq. soln.
Acetic acid, 1% v/v aq. soln.
Thermo Scientific Chemicals

Acetic acid, 1% v/v aq. soln.

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
製品番号(カタログ番号) 035569.K2
または、製品番号035569-K2
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
仕様
CAS64-19-7
ChEBICHEBI:15366
化学物質名または材質Acetic acid
濃度1% v/v aq. soln.
EINECS番号200-580-7
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only