Thermo Scientific Chemicals

Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035681.14
25 g, Each
Thermo Scientific Chemicals

Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035681.14
25 g, Each
Quantity
Catalog number: 035681.14
also known as 035681-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7787-64-6
IUPAC Name
bismuth(3+) triiodide
Molecular Formula
BiI3
InChI Key
KOECRLKKXSXCPB-UHFFFAOYSA-K
SMILES
[I-].[I-].[I-].[Bi+3]
Total Metal Impurities
0.002% max.
Particle Size
-10 Mesh
Form
Granules
Residual water
< 100 ppm
Appearance (Color)
Black

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text