Strontium chloride, ultra dry, 99.995% (metals basis)
Strontium chloride, ultra dry, 99.995% (metals basis)
Strontium chloride, ultra dry, 99.995% (metals basis)
Thermo Scientific Chemicals

Strontium chloride, ultra dry, 99.995% (metals basis)

CAS: 10476-85-4 | SrCl2
製品番号(カタログ番号) 035719.14
または、製品番号035719-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10476-85-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
さらに表示
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Particle Size-10 Mesh
Total Metal Impurities0.005% max.
Strontium chloride  is used as a source of redness in fireworks. Strontium chloride is often used as a red coloring agent in pyrotechnics. It is employed in small quantities in glass-making and metallurgy. The radioactive isotope strontium-89, used for the treatment of bone cancer, is usually administered in the form of strontium chloride. Sea water aquaria require small amounts of strontium chloride, which is consumed in the production of the exoskeletons of certain plankton. SrCl2 is useful in reducing tooth sensitivity by forming a barrier over microscopic tubules in the dentin containing nerve endings that have become exposed by gum recession. strontium chloride exposure induces parthenogenetic activation of oocytes[6] which is used in developmental biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only