Thallium(I) chloride, ultra dry, 99.999% (metals basis)
Thallium(I) chloride, ultra dry, 99.999% (metals basis)
Thallium(I) chloride, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Thallium(I) chloride, ultra dry, 99.999% (metals basis)

CAS: 7791-12-0 | ClTl | 239.83 g/mol
製品番号(カタログ番号) 035721.14
または、製品番号035721-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7791-12-0
IUPAC Nameλ¹-thallanylium chloride
Molecular FormulaClTl
InChI KeyGBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES[Cl-].[Tl+]
さらに表示
FormBeads
Total Metal Impurities0.001% max.
Appearance (Color)White
Residual water< 100 ppm
Particle Size-10 Mesh
It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only