Thallium(I) iodide, ultra dry, 99.999% (metals basis)
Thallium(I) iodide, ultra dry, 99.999% (metals basis)
Thallium(I) iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Thallium(I) iodide, ultra dry, 99.999% (metals basis)

CAS: 7790-30-9 | ITl | 331.28 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) 035722.09
または、製品番号035722-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7790-30-9
IUPAC Nameλ¹-thallanylium iodide
Molecular FormulaITl
InChI KeyCMJCEVKJYRZMIA-UHFFFAOYSA-M
SMILES[I-].[Tl+]
さらに表示
FormBeads
Total Metal Impurities0.001% max.
Residual water< 100 ppm
Particle Size-10 Mesh
Appearance (Color)Yellow
Thallium(I) iodide is used in mercury lamp to enhance its performance. It is mainly used for underwater lighting, since the blue green light produced has least absorption in water. Scintillator made by using iodide salt of thallium, sodium and cesium is utilized for radiation scintillation detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only