Europium(III) chloride, ultra dry, 99.99% (REO)
Europium(III) chloride, ultra dry, 99.99% (REO)
Europium(III) chloride, ultra dry, 99.99% (REO)
Thermo Scientific Chemicals

Europium(III) chloride, ultra dry, 99.99% (REO)

CAS: 10025-76-0 | Cl3Eu | 258.31 g/mol
製品番号(カタログ番号) 035738.03
または、製品番号035738-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10025-76-0
IUPAC Nameeuropium(3+) trichloride
Molecular FormulaCl3Eu
InChI KeyNNMXSTWQJRPBJZ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Eu+3]
さらに表示
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Appearance (Color)Yellow
Residual water< 100 ppm
Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only