Erbium(III) chloride, ultra dry, 99.9% (REO)
Erbium(III) chloride, ultra dry, 99.9% (REO)
Erbium(III) chloride, ultra dry, 99.9% (REO)
Thermo Scientific Chemicals

Erbium(III) chloride, ultra dry, 99.9% (REO)

CAS: 10138-41-7 | Cl3Er | 273.609 g/mol
製品番号(カタログ番号) 035739.06
または、製品番号035739-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10138-41-7
IUPAC Nameerbium(3+) trichloride
Molecular FormulaCl3Er
InChI KeyHDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Er+3]
さらに表示
Residual water< 100 ppm
Appearance (Color)Pink
FormColor
Total Rare Earth Oxide Impurities0.1% max.
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only