Boron tribromide, 99% min
Boron tribromide, 99% min
Boron tribromide, 99% min
Boron tribromide, 99% min
Thermo Scientific Chemicals

Boron tribromide, 99% min

As a catalyst in the manufacturing of diborane, ultra pure boron | CAS: 10294-33-4 | BBr3 | 250.52 g/mol
製品番号(カタログ番号) 035846.30
または、製品番号035846-30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10294-33-4
IUPAC Nametribromoborane
Molecular FormulaBBr3
InChI KeyILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILESBrB(Br)Br
さらに表示
Assay (unspecified)98.5% min
As a catalyst in the manufacturing of diborane, ultra pure boronBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only