Barium carbonate, ACS, 99.0-101.0%
Barium carbonate, ACS, 99.0-101.0%
Barium carbonate, ACS, 99.0-101.0%
Thermo Scientific Chemicals

Barium carbonate, ACS, 99.0-101.0%

CAS: 513-77-9 | CBaO3 | 197.34 g/mol
数量:
500 g
1 kg
2 x 1 kg
製品番号(カタログ番号) 036236.A1
または、製品番号036236-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
さらに表示
Impurity contentNa 0.02%
Impurity contentOxidizing substances (as NO₃) 0.005%
Impurity contentS 0.001%
Impurity contentSr 0.7%
Trace Metals(as Pb) 0.001%
さらに表示
Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only