Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
数量:
50 g
250 g
1 kg
5 x 1 kg
製品番号(カタログ番号) 036291.30
または、製品番号036291-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7784-27-2
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Impurity contentFe 0.002%
Assay (unspecified)98.0-102.0% Al(NO₃)₃?9H₂O
Impurity contentCa 0.005%
Impurity contentMg 0.001%
Impurity contentSO₄ 0.005%
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only