Gallium(III) oxide, 99.995% (metals basis)
Gallium(III) oxide, 99.995% (metals basis)
Gallium(III) oxide, 99.995% (metals basis)
Thermo Scientific Chemicals

Gallium(III) oxide, 99.995% (metals basis)

For vacuum deposition | CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol
製品番号(カタログ番号) 036318.06
または、製品番号036318-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12024-21-4
IUPAC Namedigallium(3+) trioxidandiide
Molecular FormulaGa2O3
InChI KeyAJNVQOSZGJRYEI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ga+3].[Ga+3]
さらに表示
Total Metal Impurities0.005% max.
For vacuum deposition. In the production of catalysts. Used as part of the manufacturing of semiconductor devices. Has been shown to demonstrate catalytic properties and has also been used as an insulating barrier in tight junctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only