Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS
Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS

CAS: 10329-15-4 | C26H26N8O2 | 482.55 g/mol
製品番号(カタログ番号) 036442.18
または、製品番号036442-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10329-15-4
IUPAC Name1,3-bis(phenylamino)urea; 3-(phenylamino)-1-(phenylimino)urea
Molecular FormulaC26H26N8O2
InChI KeyWDJPBIRJSVMEKJ-UHFFFAOYSA-N
SMILESO=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1.O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
さらに表示
Residue on ignition0.1%
Solution Testin acetone P.T.
CommentSensitivity P.T.
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide is used as an indicator in titrating iron and for the colorimetric determination of chromium and the detection of cadmium, mercury, magnesium, aldehydes, and emetine, a color developing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only