Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid
Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid
Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid
Thermo Scientific Chemicals

Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid

CAS: 10034-85-2 | HI
数量:
100 mL
500 mL
製品番号(カタログ番号) 036490.AE
または、製品番号036490-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
さらに表示
Impurity contentSO₄ 0.005%
CommentStabilizer (H₃PO₂) 1.5%
Impurity contentFe 0.001%
Assay (unspecified)47.0% HI min
Impurity contentChloride and bromide (as Cl) 0.05%
さらに表示
Hydriodic acid is used as a catalyst in the manufacture of acetic acid, as a pharmaceutical intermediate, in various disinfectants and in the preparation of organic and inorganic iodides. It is also used to prepare alkyl iodides from primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon