Thermo Scientific Chemicals

Potassium chlorate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036494.A3
2 kg, Each
Thermo Scientific Chemicals

Potassium chlorate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036494.A3
2 kg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036494.A3
also known as 036494-A3
Price (USD)
Price: 318.00
Your price:
Quantity
-

Chemical Identifiers

CAS
3811-04-9
IUPAC Name
potassium chlorate
Molecular Formula
ClKO3
InChI Key
VKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES
[K+].[O-][Cl](=O)=O
Trace Metals
(as Pb) 5ppm
Impurity content
BrO₃ 0.015%
Impurity content
SO₄ P.T. (limit about 0.002%)
Assay (unspecified)
99.0% KCLO₃ min
Impurity content
Cl 0.001%

Description

It is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text