Gallium(III) isopropoxide, mixture of oligomers, 99%
Gallium(III) isopropoxide, mixture of oligomers, 99%
Gallium(III) isopropoxide, mixture of oligomers, 99%
Thermo Scientific Chemicals

Gallium(III) isopropoxide, mixture of oligomers, 99%

CAS: 4452-61-3 | C3H8GaO | 129.819 g/mol
製品番号(カタログ番号) 036583.03
または、製品番号036583-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4452-61-3
IUPAC Nametris(propan-2-ol) gallium
Molecular FormulaC9H24GaO3
InChI KeyFUJNSPPLSODXIC-UHFFFAOYSA-N
SMILES[Ga].CC(C)O.CC(C)O.CC(C)O
さらに表示
FormViscous liquid/solid
Appearance (Color)Yellow
Assay from Supplier's CofA≥98.5%
Chloride content≤1.0%
Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only