Thermo Scientific Chemicals

Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals

Catalog number: 036731.A3
2 kg, Each
Thermo Scientific Chemicals

Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals

Catalog number: 036731.A3
2 kg, Each
Quantity
Catalog number: 036731.A3
also known as 036731-A3
Price (USD)
Price: 126.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12167-74-7
IUPAC Name
tricalcium diphosphate
Molecular Formula
Ca3O8P2
InChI Key
QORWJWZARLRLPR-UHFFFAOYSA-H
SMILES
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Assay (unspecified)
34-40% Ca

Description

Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text