Glassy carbon spherical powder, 0.4-12 micron, type 1
Glassy carbon spherical powder, 0.4-12 micron, type 1
Glassy carbon spherical powder, 0.4-12 micron, type 1
Thermo Scientific Chemicals
Glassy carbon spherical powder, 0.4-12 micron, type 1
Widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry. | CAS: 7440-44-0 | C | 12.01 g/mol
製品番号(カタログ番号) 038004.30
または、製品番号038004-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
仕様
CAS7440-44-0
化学物質名または材質Glassy carbon spherical powder
DOT情報Hazard Class: 4.1; Packaging Group: III
健康被害1H228
健康被害3P210-P240-P241-P280-P370+P378q
さらに表示
Glassy carbon spherical powder is widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
What is the difference between type 1 glassy carbon and type 2 glassy carbon?
Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.