Glassy carbon rod, 5mm (0.2in) dia, type 1
Glassy carbon rod, 5mm (0.2in) dia, type 1
Glassy carbon rod, 5mm (0.2in) dia, type 1
Thermo Scientific Chemicals

Glassy carbon rod, 5mm (0.2in) dia, type 1

Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction. | CAS: 7440-44-0
数量:
50 mm
100 mm
200 mm
製品番号(カタログ番号) 038011.DJ
または、製品番号038011-DJ
価格(JPY)
-
数量:
50 mm
一括またはカスタム形式をリクエストする
仕様
CAS7440-44-0
化学物質名または材質Glassy carbon rod
EINECS番号231-153-3
MDL番号MFCD00133992
パッケージTube
さらに表示
Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

よくあるご質問(FAQ)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.