Thermo Scientific Chemicals

Palladium, 20% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Catalog number: 038308.MD
250 mg, Each
Thermo Scientific Chemicals

Palladium, 20% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Catalog number: 038308.MD
250 mg, Each
Quantity
Catalog number: 038308.MD
also known as 038308-MD
Price (USD)
Price: 39.90
Online price: 33.65
Your price:
Quantity
-
Assay (unspecified)
Moisture, Wt% 40.0 to 65.0
Form
Powder
Assay (by ICP)
Palladium, Wt% (dry basis) 19.20 to 20.40
Appearance (Color)
Black

Description

Palladium is used as catalysis. Palladium-on-carbon has also been used as the palladium(0) catalyst in the Suzuki reaction and Stille reaction. It is also used in jewelry, dentistry, watch making, blood sugar test strips, aircraft spark plugs and in the production of surgical instruments and electrical contacts. It is also used to make professional transverse flutes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text