Indium foil, 0.05mm (0.002in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Indium foil, 0.05mm (0.002in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 038371.GH
also known as 038371-GH
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
577.00 
Save 86.35 (15%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
577.00 
Save 86.35 (15%)
Add to cart
Indium foil, 0.05mm (0.002in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number038371.GH
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
577.00 
Save 86.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only