Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides
Thermo Scientific Chemicals

Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides

CAS: 7647-14-5 | ClNa | 58.44 g/mol
製品番号(カタログ番号)数量
038473.KT1 Ea.
製品番号(カタログ番号) 038473.KT
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
仕様
CAS7647-14-5
ChEBICHEBI:26710
化学物質名または材質Sodium chloride Crystal Optic Rectangle
EINECS番号231-598-3
IUPAC名sodium chloride
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only