Silicon granular, virgin poly fines, 99.999% (metals basis)
Silicon granular, virgin poly fines, 99.999% (metals basis)
Silicon granular, virgin poly fines, 99.999% (metals basis)
Thermo Scientific Chemicals

Silicon granular, virgin poly fines, 99.999% (metals basis)

CAS: 7440-21-3 | Si | 28.09 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 038542.36
または、製品番号038542-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
さらに表示
FormChunks to powder
Particle Size~1/4in and down
Total Metal Impurities≤ 0.001%
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only