Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL

Used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry. | CAS: 11145-27-0 | AlSi | 55.067 g/mol
数量:
50 mL
100 mL
500 mL
製品番号(カタログ番号) 038721.AD
または、製品番号038721-AD
価格(JPY)
-
数量:
50 mL
一括またはカスタム形式をリクエストする
仕様
CAS11145-27-0
化学物質名または材質Aluminum
DOT情報Hazard Class: 8
EINECS番号231-714-2
健康被害1H290-H314-H335
さらに表示
Aluminum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only