Thermo Scientific Chemicals
Glassy Carbon Boats
Catalog number: 039026.KT
29 x 16 x 10mm, 2 mL, Each
Thermo Scientific Chemicals
Glassy Carbon Boats
Catalog number: 039026.KT
29 x 16 x 10mm, 2 mL, Each
Dimensions (L x W x H)
Capacity (Metric)
Catalog number: 039026.KT
also known as 039026-KT
Price (USD)
Price: 230.00
Online price: 195.65
Your price:
Quantity
-
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Resistance to all wet decomposition agents and abrasive wear
  • No memory effects or contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistance to thermal shock allows rapid heating and cooling times
  • Good electrical conductivity
  • Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal­ growing such as doped halogenide crystals