Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)

Form: Approx. 3.3g/25 x 25mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
数量:
25 x 25 mm
50 x 50 mm
100 x 100 mm
200 x 200 mm
製品番号(カタログ番号) 039181.FI
または、製品番号039181-FI
価格(JPY)
-
数量:
50 x 50 mm
一括またはカスタム形式をリクエストする
仕様
外見Silver-colored foil
定量パーセント範囲99.9% (Metals basis)
CAS7440-22-4
ChEBICHEBI:9141
化学物質名または材質Silver
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only