n-Butyl acetate, HPLC Grade, 99.5+%
n-Butyl acetate, HPLC Grade, 99.5+%
n-Butyl acetate, HPLC Grade, 99.5+%
Thermo Scientific Chemicals

n-Butyl acetate, HPLC Grade, 99.5+%

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
数量:
250 mL
1 L
4 L
4 x 1 L
製品番号(カタログ番号) 039197.AK
または、製品番号039197-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
さらに表示
Optical Absorbance at254 nm: ≤1.00 AU
Optical Absorbance at259 nm: ≤0.20
Optical Absorbance at300 nm: ≤0.02 AU
Optical Absorbance at400 nm: ≤0.01 AU
Optical Absorbance at275 nm: ≤0.04 AU
さらに表示
n-Butyl acetate is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only