Caffeine, 99.7%
Caffeine, 99.7%
Caffeine, 99.7%
Thermo Scientific Chemicals

Caffeine, 99.7%

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 039214.14
または、製品番号039214-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS58-08-2
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
さらに表示
Residue on ignition0.1%, max
Loss on Drying0.5%, max
Appearance (Color)White to off-white
Assay (Titration)>/= 99.6% ( on dry basis)
FormPowder
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only