Thermo Scientific Chemicals

Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals

Catalog number: 039215.22
100 g, Each
Thermo Scientific Chemicals

Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals

Catalog number: 039215.22
100 g, Each
Quantity
Catalog number: 039215.22
also known as 039215-22
Price (USD)
Price: 151.00
Online price: 128.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13454-71-2
IUPAC Name
cerium(3+) phosphate
Molecular Formula
CeO4P
InChI Key
TYAVIWGEVOBWDZ-UHFFFAOYSA-K
SMILES
[Ce+3].[O-]P([O-])([O-])=O
Assay (unspecified)
98.6% min

Description

Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text