Thermo Scientific Chemicals

Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039216.36
500 g, Each
Thermo Scientific Chemicals

Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039216.36
500 g, Each
Quantity
Catalog number: 039216.36
also known as 039216-36
Price (USD)

Chemical Identifiers

CAS
53169-24-7
IUPAC Name
zirconium(4+) cerium(3+) oxidandiide
Molecular Formula
CeOZr
InChI Key
WPQBNJRIWONKBL-UHFFFAOYSA-N
SMILES
[O--].[Zr+4].[Ce+3]
Total Metal Impurities
0.5% max.

Description

Used as wear resistant grinding and dispersion media. Used in fuel cell materials (SOFC), body armors. Also finds applications in catalytic converter, honey comb, precision ceramic parts, abrasive wheels, oxygen sensors, cutting tools. Used as ceramic steel. Other uses include screen printing, dip and spin coating and APS coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text