Tris(cyclopentadienyl)neodymium(III), 99.5%
Tris(cyclopentadienyl)neodymium(III), 99.5%
Tris(cyclopentadienyl)neodymium(III), 99.5%
Thermo Scientific Chemicals

Tris(cyclopentadienyl)neodymium(III), 99.5%

CAS: 1273-98-9 | C15H15Nd | 339.527 g/mol
製品番号(カタログ番号) 039300.03
または、製品番号039300-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS1273-98-9
化学物質名または材質Tris(cyclopentadienyl)neodymium(III)
DOT情報Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECS番号215-057-9
式量339.53
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only