Magnesium pyrophosphate
Magnesium pyrophosphate
Magnesium pyrophosphate
Magnesium pyrophosphate
Thermo Scientific Chemicals

Magnesium pyrophosphate

CAS: 13446-24-7 | Mg2O7P2 | 222.551 g/mol
数量:
50 g
250 g
1 kg
製品番号(カタログ番号) 039317.A1
または、製品番号039317-A1
価格(JPY)
-
見積もりを依頼する
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13446-24-7
IUPAC Namedimagnesium(2+) (phosphonatooxy)phosphonate
Molecular FormulaMg2O7P2
InChI KeyXZTWHWHGBBCSMX-UHFFFAOYSA-J
SMILES[Mg++].[Mg++].[O-]P([O-])(=O)OP([O-])([O-])=O
さらに表示
Assay (unspecified)21% Mg min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only