Carbonylchlorobis(triphenylphosphine)iridium(I)
Carbonylchlorobis(triphenylphosphine)iridium(I)
Carbonylchlorobis(triphenylphosphine)iridium(I)
Thermo Scientific Chemicals

Carbonylchlorobis(triphenylphosphine)iridium(I)

CAS: 14871-41-1 | C37H30ClIrOP2 | 780.26 g/mol
製品番号(カタログ番号) 039413.01
または、製品番号039413-01
価格(JPY)
-
数量:
0.25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14871-41-1
IUPAC Namechloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane)
Molecular FormulaC37H30ClIrOP2
InChI KeyPEPSDKMVHYXIMJ-UHFFFAOYSA-M
SMILESCl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Assay (unspecified)Ir 24% min
Carbonylchlorobis(triphenylphosphine)iridium(I) is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only